This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02240507_g1
See other BB GE Assays ›
Gene Symbol:
BB
Gene Name:
RING/U-box superfamily protein
Gene Aliases:
AT3G63530, BB2, BIG BROTHER, ENHANCER1 OF DA1, EOD1
Chromosome Location:
Chr.3: 23456055 - 23458732 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02240507_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035845.2 NP_001030922.1 4 - 5 913 74
NM_148885.3 NP_680148.1 4 - 5 637 74
GenBank mRNA AK227673.1 - 4 - 5 634 74
BT010197.1 - 2 - 3 384 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell proliferation
protein ubiquitination
negative regulation of organ growth
floral organ development
protein autoubiquitination
positive regulation of leaf senescence
ubiquitin-protein transferase activity
protein binding
ubiquitin conjugating enzyme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0