This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02240297_g1
See other AT3G29638 GE Assays ›
Gene Symbol:
AT3G29638
Gene Name:
General transcription factor 2-related zinc finger protein
Gene Aliases:
AT3G29638
Chromosome Location:
Chr.3: 11473109 - 11475559 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02240297_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G29638
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_148767.2 NP_683609.2 1 - 2 684 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0