This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02239665_g1
See other CIP7 GE Assays ›
Gene Symbol:
CIP7
Gene Name:
COP1-interacting protein 7
Gene Aliases:
AT4G27430, COP1-interacting protein 7, F27G19.30, F27G19_30
Chromosome Location:
Chr.4: 13718007 - 13723461 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02239665_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIP7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001125596.2 NP_001119068.1 7 - 8 967 61
NM_118877.4 NP_194473.1 7 - 8 1639 61
GenBank mRNA AB012912.1 - 7 - 8 967 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to light stimulus
anthocyanin-containing compound biosynthetic process
chlorophyll biosynthetic process
positive regulation of transcription, DNA-templated
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0