This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02237701_g1
See other ZIM GE Assays ›
Gene Symbol:
ZIM
Gene Name:
GATA-type zinc finger protein with TIFY domain-containing protein
Gene Aliases:
AT4G24470, AtTIFY1, GATA TRANSCRIPTION FACTOR 25, GATA25, T22A6.300, T22A6_300, TIFY1, Zinc-finger protein expressed in Inflorescence Meristem
Chromosome Location:
Chr.4: 12645542 - 12647985 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02237701_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZIM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203892.1 NP_001190821.1 4 - 5 856 87
NM_118580.4 NP_194178.1 4 - 5 876 87
NM_179104.3 NP_849435.1 5 - 6 758 87
GenBank mRNA AY086038.1 - 4 - 5 774 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
circadian rhythm
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0