This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02237406_g1
See other NLP7 GE Assays ›
Gene Symbol:
NLP7
Gene Name:
NIN like protein 7
Gene Aliases:
AT4G24020, NIN like protein 7, T19F6.10, T19F6_10
Chromosome Location:
Chr.4: 12479528 - 12484049 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02237406_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NLP7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001341638.1 NP_001328405.1 1 - 2 703 92
NM_118534.6 NP_194133.1 1 - 2 703 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001341638.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to water deprivation
stomatal movement
response to nitrate
nitrate assimilation
transcription regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0