This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02236412_g1
See other ALDH22A1 GE Assays ›
Gene Symbol:
ALDH22A1
Gene Name:
aldehyde dehydrogenase 22A1
Gene Aliases:
AT3G66658, T8E24.4, T8E24_4, aldehyde dehydrogenase 22A1
Chromosome Location:
Chr.3: 2094998 - 2099250 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02236412_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH22A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111545.3 NP_187321.2 7 - 8 893 106
NM_202513.3 NP_974242.1 7 - 8 1006 106
GenBank mRNA AJ584646.1 - 7 - 8 776 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

cellular aldehyde metabolic process
aldehyde dehydrogenase (NAD) activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (non-phosphorylating) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0