This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02236101_g1
See other MEE32 GE Assays ›
Gene Symbol:
MEE32
Gene Name:
dehydroquinate dehydratase, putative / shikimate dehydrogenase
Gene Aliases:
AT3G06350, EMB3004, EMBRYO DEFECTIVE 3004, F24P17.18, F24P17_18, MATERNAL EFFECT EMBRYO ARREST 32
Chromosome Location:
Chr.3: 1924270 - 1927850 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02236101_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MEE32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111510.4 NP_187286.1 2 - 3 616 144

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

chorismate biosynthetic process
embryo development ending in seed dormancy
shikimate metabolic process
3-dehydroquinate dehydratase activity
shikimate 3-dehydrogenase (NADP+) activity
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0