This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02234911_g1
See other TPR10 GE Assays ›
Gene Symbol:
TPR10
Gene Name:
ankyrin repeat family protein
Gene Aliases:
AT3G04710, F7O18.18, F7O18_18, tetratricopeptide repeat 10
Chromosome Location:
Chr.3: 1276668 - 1281170 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02234911_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPR10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035558.1 NP_001030635.1 10 - 11 1202 108
NM_001202879.1 NP_001189808.1 12 - 13 2012 108
NM_111343.3 NP_187122.1 10 - 11 1261 108
GenBank mRNA AK175477.1 - 10 - 11 1205 108
AY084331.1 - 10 - 11 108
AY136361.1 - 10 - 11 1209 108
BT000201.1 - 10 - 11 1060 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0