This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02231388_g1
See other ACD1 GE Assays ›
Gene Symbol:
ACD1
Gene Name:
Pheophorbide a oxygenase family protein with Rieske 2Fe-2S domain-containing protein
Gene Aliases:
ACCELERATED CELL DEATH 1, AT3G44880, LETHAL LEAF-SPOT 1 HOMOLOG, LLS1, PAO, PHEOPHORBIDE A OXYGENASE
Chromosome Location:
Chr.3: 16383595 - 16386344 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02231388_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_114357.6 NP_190074.1 6 - 7 1623 82
GenBank mRNA AY093092.1 - 6 - 7 1408 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

cell death
defense response to bacterium, incompatible interaction
flower development
fruit development
chlorophyll catabolic process
chlorophyllide a oxygenase [overall] activity
pheophorbide a oxygenase activity
iron-sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0