This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02230655_g1
See other ATMIN7 GE Assays ›
Gene Symbol:
ATMIN7
Gene Name:
HOPM interactor 7
Gene Aliases:
AT3G43300, BEN1, BFA-VISUALIZED ENDOCYTIC TRAFFICKING DEFECTIVE1, HOPM interactor 7, MIN7
Chromosome Location:
Chr.3: 15233845 - 15245281 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02230655_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATMIN7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203077.1 NP_001190006.1 7 - 8 819 108
NM_001339097.1 NP_001327198.1 7 - 8 967 108
NM_114198.4 NP_189916.4 7 - 8 720 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

immune response
vesicle-mediated transport
regulation of ARF protein signal transduction
defense response to bacterium
innate immune response
ARF guanyl-nucleotide exchange factor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0