This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02228696_g1
See other PS2 GE Assays ›
Gene Symbol:
PS2
Gene Name:
inorganic pyrophosphatase 1
Gene Aliases:
AT1G73010, ATPS2, AtPPsPase1, F3N23.21, F3N23_21, PPsPase1, phosphate starvation-induced gene 2, pyrophosphate-specific phosphatase1
Chromosome Location:
Chr.1: 27464528 - 27466348 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02228696_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105959.3 NP_565052.1 1 - 2 335 78
GenBank mRNA AK176548.1 - 1 - 2 237 78
BT030334.1 - 1 - 2 167 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

cellular response to phosphate starvation
protein tetramerization
cellular response to hypoxia
inorganic diphosphatase activity
pyrophosphatase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0