This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02228348_g1
See other JAZ6 GE Assays ›
Gene Symbol:
JAZ6
Gene Name:
jasmonate-zim-domain protein 6
Gene Aliases:
AT1G72450, T10D10.8, T10D10_8, TIFY DOMAIN PROTEIN 11B, TIFY11B, jasmonate-zim-domain protein 6
Chromosome Location:
Chr.1: 27273968 - 27276562 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02228348_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: JAZ6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334533.1 NP_001321693.1 2 - 3 458 87
NM_105904.4 NP_565043.1 2 - 3 703 87
GenBank mRNA AF370147.1 - 2 - 3 374 87
AY059089.1 - 2 - 3 277 87
AY088035.1 - 2 - 3 429 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to wounding
response to jasmonic acid
regulation of defense response
regulation of jasmonic acid mediated signaling pathway
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0