This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02227591_g1
See other RAD52-1 GE Assays ›
Gene Symbol:
RAD52-1
Gene Name:
cobalt ion binding protein
Gene Aliases:
AT1G71310, F3I17.4, F3I17_4, ODB1, Organellar DNA-Binding protein 1, RAD52-1A, RAD52-1B.1, RAD52-1B.2, radiation sensitive 51-1
Chromosome Location:
Chr.1: 26877745 - 26880064 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02227591_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAD52-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105800.2 NP_177287.1 4 - 5 716 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

Group II intron splicing
double-strand break repair via homologous recombination
DNA repair
DNA binding
double-stranded DNA binding
single-stranded DNA binding
protein binding
cobalt ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0