This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02227141_g1
See other TIFY7 GE Assays ›
Gene Symbol:
TIFY7
Gene Name:
TIFY domain/Divergent CCT motif family protein
Gene Aliases:
AT1G70700, F5A18.12, F5A18_12, JASMONATE-ZIM-DOMAIN PROTEIN 9, JAZ9
Chromosome Location:
Chr.1: 26654529 - 26657350 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02227141_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIFY7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001036187.5 NP_001031264.4 4 - 5 598 72
NM_001334477.1 NP_001320988.1 4 - 5 598 72
NM_105738.8 NP_177227.5 4 - 5 837 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to wounding
response to jasmonic acid
regulation of defense response
regulation of jasmonic acid mediated signaling pathway
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0