This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02227033_g1
See other CRK3 GE Assays ›
Gene Symbol:
CRK3
Gene Name:
cysteine-rich RLK (RECEPTOR-like protein kinase) 3
Gene Aliases:
AT1G70530, F24J13.10, F24J13_10, cysteine-rich RLK (RECEPTOR-like protein kinase) 3
Chromosome Location:
Chr.1: 26588391 - 26591776 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02227033_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334460.1 NP_001320922.1 5 - 6 1554 139
NM_001334461.1 NP_001320921.1 5 - 6 1610 139
NM_105721.2 NP_177210.1 5 - 6 1615 139

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein autophosphorylation
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0