This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02227028_g1
See other CRK2 GE Assays ›
Gene Symbol:
CRK2
Gene Name:
cysteine-rich RLK (RECEPTOR-like protein kinase) 2
Gene Aliases:
ALTERED SEED GERMINATION 6, ASG6, AT1G70520, F24J13.9, F24J13_9, cysteine-rich RLK (RECEPTOR-like protein kinase) 2
Chromosome Location:
Chr.1: 26584729 - 26587628 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02227028_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105720.4 NP_177209.1 6 - 7 1909 89
GenBank mRNA AK118924.1 - 6 - 7 1700 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

response to ozone
protein autophosphorylation
reactive oxygen species metabolic process
protein serine/threonine kinase activity
protein binding
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0