This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02226434_g1
See other CM3 GE Assays ›
Gene Symbol:
CM3
Gene Name:
chorismate mutase 3
Gene Aliases:
AT1G69370, AtCM3, F10D13.6, F10D13_6, chorismate mutase 3, cm-3
Chromosome Location:
Chr.1: 26079956 - 26081727 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02226434_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105604.2 NP_177096.1 2 - 3 537 116
GenBank mRNA AK117860.1 - 2 - 3 411 116
BT005306.1 - 2 - 3 395 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

tryptophan biosynthetic process
aromatic amino acid family biosynthetic process
defense response to bacterium
chorismate metabolic process
chorismate mutase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm