This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02225442_g1
See other ARR11 GE Assays ›
Gene Symbol:
ARR11
Gene Name:
response regulator 11
Gene Aliases:
AT1G67710, F12A21.15, F12A21_15, response regulator 11
Chromosome Location:
Chr.1: 25376679 - 25379217 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02225442_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARR11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334321.1 NP_001320475.1 1 - 2 662 82
NM_105439.3 NP_176938.1 2 - 3 581 82
GenBank mRNA AK228216.1 - 2 - 3 360 82
BT008768.1 - 2 - 3 269 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

phosphorelay signal transduction system
response to cytokinin
cytokinin-activated signaling pathway
regulation of abscisic acid-activated signaling pathway
regulation of root meristem growth
regulation of jasmonic acid mediated signaling pathway
regulation of salicylic acid mediated signaling pathway
phosphorelay response regulator activity
transcription factor activity, sequence-specific DNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0