This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02224748_g1
See other LIG6 GE Assays ›
Gene Symbol:
LIG6
Gene Name:
DNA LIGASE 6
Gene Aliases:
AT1G66730, AtLIG6, DNA LIGASE 6, F4N21.14, F4N21_14
Chromosome Location:
Chr.1: 24884681 - 24892046 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02224748_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIG6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_105343.3 NP_176845.2 5 - 6 2982 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA ligase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

response to molecule of bacterial origin
DNA ligation
lagging strand elongation
DNA repair
double-strand break repair via nonhomologous end joining
response to oxidative stress
response to cold
seed germination
response to UV-C
DNA integration
seed development
DNA ligation involved in DNA repair
DNA biosynthetic process
response to bleomycin
positive regulation of cellular response to X-ray
DNA binding
DNA ligase (ATP) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0