This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02223129_gH
See other SSP4B GE Assays ›
Gene Symbol:
SSP4b
Gene Name:
SCP1-like small phosphatase 4b
Gene Aliases:
AT4G18140, F15J5.110, F15J5_110, SCP1-like small phosphatase 4b
Chromosome Location:
Chr.4: 10046287 - 10049438 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02223129_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSP4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001036584.3 NP_001031661.2 5 - 6 1291 125
NM_001203831.1 NP_001190760.1 5 - 6 1291 125
NM_001341261.1 NP_001328767.1 5 - 6 1332 125
NM_001341262.1 NP_001328766.1 5 - 6 1291 125
NM_001341263.1 NP_001328768.1 4 - 5 1558 125
NM_117924.7 NP_193548.7 4 - 5 1514 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
phosphoprotein phosphatase activity
CTD phosphatase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0