This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02221413_g1
See other CLA1 GE Assays ›
Gene Symbol:
CLA1
Gene Name:
Deoxyxylulose-5-phosphate synthase
Gene Aliases:
1-DEOXY-D-XYLULOSE 5-PHOSPHATE SYNTHASE, 1-DEOXY-D-XYLULOSE 5-PHOSPHATE SYNTHASE 2, 1-deoxy-D-xylulose 5-phosphate (DXP) synthase 1, AT4G15560, AtCLA1, CLA, CLOROPLASTOS ALTERADOS 1, DEF, DL3821W, DXPS2, DXS, DXS1, FCAALL.24
Chromosome Location:
Chr.4: 8883825 - 8887667 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02221413_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117647.3 NP_193291.1 1 - 2 544 74
GenBank mRNA BT002340.1 - 1 - 2 151 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

chlorophyll biosynthetic process
terpenoid biosynthetic process
isopentenyl diphosphate biosynthetic process, methylerythritol 4-phosphate pathway
1-deoxy-D-xylulose 5-phosphate biosynthetic process
protein binding
1-deoxy-D-xylulose-5-phosphate synthase activity
transferase activity, transferring aldehyde or ketonic groups
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0