This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02221299_m1
See other AT4G15470 GE Assays ›
Gene Symbol:
AT4G15470
Gene Name:
Bax inhibitor-1 family protein
Gene Aliases:
AT4G15470, AtLFG5, DL3775W, FCAALL.58, LFG5, LIFEGUARD 5
Chromosome Location:
Chr.4: 8843519 - 8845810 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02221299_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G15470
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117636.3 NP_567466.1 1 - 2 440 73
GenBank mRNA AY050443.1 - 1 - 2 343 73
AY097356.1 - 1 - 2 298 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0