This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02220847_g1
See other AT4G14815 GE Assays ›
Gene Symbol:
AT4G14815
Gene Name:
Bifunctional inhibitor/lipid-transfer protein/seed storage 2S albumin superfamily protein
Gene Aliases:
AT4G14815
Chromosome Location:
Chr.4: 8505043 - 8505952 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02220847_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G14815
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117567.4 NP_567445.1 2 - 3 412 120
GenBank mRNA AB246333.1 - 2 - 3 333 120
DQ446836.1 - 2 - 3 333 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header