This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02220586_s1
See other IBR10 GE Assays ›
Gene Symbol:
IBR10
Gene Name:
indole-3-butyric acid response 10
Gene Aliases:
ARABIDOPSIS THALIANA DELTA(3), AT4G14430, ATECI2, DELTA(2)-ENOYL COA ISOMERASE 2, DELTA(3), DL3255C, ECHIB, ECI2, ENOYL-COA HYDRATASE/ISOMERASE B, FCAALL.43, PEC12, indole-3-butyric acid response 10
Chromosome Location:
Chr.4: 8304442 - 8305857 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02220586_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IBR10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117522.3 NP_193179.1 1 1220 69
GenBank mRNA AF380640.1 - 1 1052 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
fatty acid catabolic process
root hair elongation
indolebutyric acid metabolic process
response to indolebutyric acid
dodecenoyl-CoA delta-isomerase activity
enoyl-CoA hydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0