This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02220504_gH
See other CKI1 GE Assays ›
Gene Symbol:
CKI1
Gene Name:
casein kinase I
Gene Aliases:
AT4G14340, CASEIN KINASE I-LIKE 11, CKL11, DL3210C, FCAALL.68, casein kinase I
Chromosome Location:
Chr.4: 8248160 - 8251998 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02220504_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CKI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340918.1 NP_001328596.1 6 - 7 854 72
NM_117511.5 NP_193170.1 6 - 7 854 72
GenBank mRNA AF250343.1 - 6 - 7 678 72
AY052364.1 - 6 - 7 824 72
AY060528.1 - 6 - 7 524 72
AY943850.1 - 6 - 7 524 72
X78818.1 - 695 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
protein autophosphorylation
protein serine/threonine kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0