This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02219893_g1
See other IIL1 GE Assays ›
Gene Symbol:
IIL1
Gene Name:
isopropyl malate isomerase large subunit 1
Gene Aliases:
AT4G13430, ATLEUC1, T9E8.170, T9E8_170, isopropyl malate isomerase large subunit 1
Chromosome Location:
Chr.4: 7803863 - 7807984 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02219893_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IIL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_117417.4 NP_567405.1 11 - 12 1250 112
GenBank mRNA AY045842.1 - 11 - 12 1124 112
AY117151.1 - 11 - 12 1055 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

leucine biosynthetic process
glucosinolate biosynthetic process
response to cadmium ion
3-isopropylmalate dehydratase activity
intramolecular transferase activity, transferring hydroxy groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0