This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02216391_g1
See other SCO1 GE Assays ›
Gene Symbol:
SCO1
Gene Name:
Translation elongation factor EFG/EF2 protein
Gene Aliases:
AT1G62750, ATSCO1, ATSCO1/CPEF-G, F23N19.11, F23N19_11, SNOWY COTYLEDON 1
Chromosome Location:
Chr.1: 23233434 - 23236447 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02216391_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104952.2 NP_564801.1 2 - 3 1793 102
GenBank mRNA AY142646.1 - 2 - 3 1667 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor translation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
chloroplast organization
post-embryonic development
seed germination
mitochondrial translation
ribosome disassembly
mRNA binding
GTPase activity
ATP binding
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0