This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02216256_m1
See other FMO GS-OX4 GE Assays ›
Gene Symbol:
FMO GS-OX4
Gene Name:
flavin-monooxygenase glucosinolate S-oxygenase 4
Gene Aliases:
AT1G62570, T3P18.13, T3P18_13, flavin-monooxygenase glucosinolate S-oxygenase 4
Chromosome Location:
Chr.1: 23168767 - 23172175 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02216256_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FMO GS-OX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104935.2 NP_564797.1 2 - 3 1055 68
GenBank mRNA AY054642.1 - 2 - 3 686 68
AY081554.1 - 2 - 3 615 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glucosinolate biosynthetic process
oxidation-reduction process
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding
4-methylthiopropyl glucosinolate S-oxygenase activity
8-methylthiopropyl glucosinolate S-oxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0