This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02212411_g1
See other XRN4 GE Assays ›
Gene Symbol:
XRN4
Gene Name:
exoribonuclease 4
Gene Aliases:
5'-3' EXORIBONUCLEASE, ACC INSENSITIVE 1, AIN1, AT1G54490, ATXRN4, EIN5, ETHYLENE INSENSITIVE 5, EXORIBONUCLEASE 4, F20D21.30, F20D21_30, exoribonuclease 4
Chromosome Location:
Chr.1: 20350109 - 20356916 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02212411_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XRN4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001333652.1 NP_001321327.1 12 - 13 1805 67
NM_104327.4 NP_175851.1 12 - 13 1600 67
GenBank mRNA AF286718.1 - 12 - 13 1519 67
AY064012.1 - 12 - 13 1516 67
AY091411.1 - 12 - 13 1409 67
BT026022.1 - 12 - 13 1495 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, exonucleolytic
nuclear-transcribed mRNA catabolic process
mRNA catabolic process
gravitropism
response to ethylene
unidimensional cell growth
ethylene-activated signaling pathway
response to 1-aminocyclopropane-1-carboxylic acid
phloem or xylem histogenesis
heat acclimation
miRNA catabolic process
deadenylation-independent decapping of nuclear-transcribed mRNA
regulation of gene expression, epigenetic
cell division
positive regulation of mRNA catabolic process
cellular heat acclimation
nucleic acid binding
RNA binding
mRNA binding
5'-3' exoribonuclease activity
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0