This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02211947_g1
See other SRF6 GE Assays ›
Gene Symbol:
SRF6
Gene Name:
STRUBBELIG-receptor family 6
Gene Aliases:
AT1G53730, F22G10.31, F22G10_31, STRUBBELIG-receptor family 6
Chromosome Location:
Chr.1: 20061405 - 20065713 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02211947_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084247.2 NP_001077716.1 5 - 6 804 79
NM_104251.4 NP_175777.1 5 - 6 727 79
GenBank mRNA AK227057.1 - 5 - 6 480 79
AY518291.1 - 5 - 6 508 79
BT010600.1 - 5 - 6 438 79
FJ708659.1 - 5 - 6 438 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0