This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02208250_g1
See other RSH1 GE Assays ›
Gene Symbol:
RSH1
Gene Name:
RELA/SPOT homolog 1
Gene Aliases:
AT-RSH1, AT4G02260, ATRSH1, RELA-SPOT HOMOLOG 1, RELA/SPOT HOMOLOG 1, RELA/SPOT homolog 1, T2H3.10, T2H3_10
Chromosome Location:
Chr.4: 985123 - 991506 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02208250_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RSH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340369.1 NP_001329795.1 16 - 17 1900 77
NM_116459.4 NP_567226.1 16 - 17 1917 77
NM_178956.4 NP_849287.2 16 - 17 1911 77
NM_202772.2 NP_974501.1 16 - 17 1908 77
GenBank mRNA AF225702.1 - 16 - 17 1761 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

response to wounding
leaf senescence
guanosine tetraphosphate metabolic process
photosynthesis
protein binding
GTP diphosphokinase activity
guanosine-3',5'-bis(diphosphate) 3'-diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0