This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02207305_g1
See other CIP4.1 GE Assays ›
Gene Symbol:
CIP4.1
Gene Name:
COP1-interacting protein 4.1
Gene Aliases:
AT4G00930, A_TM018A10.17, A_TM018A10_17, COP1-interacting protein 4.1, T18A10.23, T18A10_23
Chromosome Location:
Chr.4: 396444 - 400994 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02207305_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIP4.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340279.1 NP_001329938.1 5 - 6 1362 127
NM_001340280.1 NP_001329937.1 5 - 6 1362 127
NM_001340281.1 NP_001329936.1 5 - 6 1358 127
NM_001340282.1 NP_001329935.1 5 - 6 1352 127
NM_116320.6 NP_567201.5 5 - 6 1362 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0