This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02206931_g1
See other AT4G00290 GE Assays ›
Gene Symbol:
AT4G00290
Gene Name:
Mechanosensitive ion channel protein
Gene Aliases:
AT4G00290, A_IG005I10.9, A_IG005I10_9, F5I10.9, F5I10_9
Chromosome Location:
Chr.4: 122851 - 125591 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02206931_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G00290
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_116250.5 NP_567165.2 6 - 7 1499 132
GenBank mRNA AK176463.1 - 6 - 7 1499 132
AK176579.1 - 6 - 7 1499 132
AY064027.1 - 6 - 7 1497 132
AY096611.1 - 5 - 6 1380 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

cellular response to oxidative stress
mechanically-gated ion channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0