This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02205020_g1
See other ZML2 GE Assays ›
Gene Symbol:
ZML2
Gene Name:
ZIM-LIKE 2
Gene Aliases:
AT1G51600, F5D21.22, F5D21_22, GATA TRANSCRIPTION FACTOR 28, GATA28, TIFY2A, ZIM-LIKE 2
Chromosome Location:
Chr.1: 19132221 - 19135630 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02205020_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZML2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_104038.4 NP_564593.1 4 - 5 998 66
NM_202273.3 NP_974002.1 4 - 5 760 66
GenBank mRNA AB119061.1 - 4 - 5 672 66
AY150395.1 - 3 - 4 531 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0