This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02204232_g1
See other CPL3 GE Assays ›
Gene Symbol:
CPL3
Gene Name:
C-terminal domain phosphatase-like 3
Gene Aliases:
AT2G33540, ATCPL3, C-terminal domain phosphatase-like 3, F4P9.31, F4P9_31
Chromosome Location:
Chr.2: 14203401 - 14209192 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02204232_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001336459.1 NP_001325202.1 1 - 2 1282 135
NM_128914.3 NP_180912.2 2 - 3 1157 135

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001336459.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

response to salt stress
negative regulation of abscisic acid-activated signaling pathway
dephosphorylation of RNA polymerase II C-terminal domain
negative regulation of RNA interference
protein C-terminus binding
CTD phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0