This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02202663_g1
See other ASP1 GE Assays ›
Gene Symbol:
ASP1
Gene Name:
aspartate aminotransferase 1
Gene Aliases:
ASPARTATE AMINOTRANSFERASE 1, AT2G30970, F7F1.18, F7F1_18, aspartate aminotransferase 1
Chromosome Location:
Chr.2: 13178829 - 13182050 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02202663_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001124949.1 NP_001118421.1 10 - 11 1314 97
NM_128651.5 NP_180654.1 9 - 10 1403 97
GenBank mRNA AY059912.1 - 9 - 10 1271 97
AY128806.1 - 9 - 10 1220 97
U15026.1 - 9 - 10 1318 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

2-oxoglutarate metabolic process
aspartate metabolic process
glutamate metabolic process
biosynthetic process
response to cadmium ion
L-aspartate:2-oxoglutarate aminotransferase activity
copper ion binding
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header