This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02202405_g1
See other AT5G19510 GE Assays ›
Gene Symbol:
AT5G19510
Gene Name:
Translation elongation factor EF1B/ribosomal protein S6 family protein
Gene Aliases:
AT5G19510, T20D1.30, T20D1_30
Chromosome Location:
Chr.5: 6581658 - 6583248 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02202405_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G19510
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121956.3 NP_568375.2 4 - 5 501 67
GenBank mRNA AF360304.1 - 4 - 5 429 67
AJ249597.1 - 4 - 5 392 67
AY056354.1 - 4 - 5 390 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
defense response to bacterium
guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0