This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02201964_g1
See other CPN60ALPHA2 GE Assays ›
Gene Symbol:
Cpn60alpha2
Gene Name:
TCP-1/cpn60 chaperonin family protein
Gene Aliases:
AT5G18820, EMB3007, F17K4.70, F17K4_70, chaperonin-60alpha2, embryo defective 3007
Chromosome Location:
Chr.5: 6271386 - 6274599 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02201964_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpn60alpha2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001343571.1 NP_001330414.1 4 - 5 709 88
NM_001343572.1 NP_001330413.1 4 - 5 627 88
NM_121887.3 NP_197383.1 4 - 5 693 88
GenBank mRNA AK221565.1 - 4 - 5 578 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein folding
embryo development ending in seed dormancy
protein refolding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0