This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02201061_g1
See other APG5 GE Assays ›
Gene Symbol:
APG5
Gene Name:
autophagy protein Apg5 family
Gene Aliases:
AT5G17290, ATATG5, ATG5, AUTOPHAGY 5, MKP11.20, MKP11_20
Chromosome Location:
Chr.5: 5686135 - 5689947 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02201061_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APG5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121735.5 NP_197231.1 3 - 4 422 89
GenBank mRNA AK117572.1 - 3 - 4 308 89
BT005100.1 - 3 - 4 296 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s)

autophagosome assembly
mitophagy
C-terminal protein lipidation
autophagy
cellular response to nitrogen starvation
leaf senescence
response to starvation
nucleophagy
defense response to fungus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0