This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02200575_g1
See other PFA-DSP5 GE Assays ›
Gene Symbol:
PFA-DSP5
Gene Name:
Phosphotyrosine protein phosphatases superfamily protein
Gene Aliases:
AT5G16480, AtPFA-DSP5, MQK4.21, MQK4_21, plant and fungi atypical dual-specificity phosphatase 5
Chromosome Location:
Chr.5: 5381069 - 5382939 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02200575_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PFA-DSP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121653.4 NP_197152.1 3 - 4 625 89
GenBank mRNA BT025778.1 - 3 - 4 304 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

dephosphorylation
protein tyrosine phosphatase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0