This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02198748_g1
See other TRA2 GE Assays ›
Gene Symbol:
TRA2
Gene Name:
Aldolase-type TIM barrel family protein
Gene Aliases:
AT5G13420, T22N19.70, T22N19_70, transaldolase 2
Chromosome Location:
Chr.5: 4301792 - 4304315 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02198748_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_121345.3 NP_196846.1 5 - 6 995 66
GenBank mRNA AY056296.1 - 5 - 6 951 66
AY087135.1 - 5 - 6 993 66
AY114075.1 - 5 - 6 892 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
pentose-phosphate shunt
lignin biosynthetic process
shikimate biosynthetic process
response to cadmium ion
sedoheptulose-7-phosphate:D-glyceraldehyde-3-phosphate glyceronetransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0