This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02197424_gH
See other DIN2 GE Assays ›
Gene Symbol:
DIN2
Gene Name:
Glycosyl hydrolase superfamily protein
Gene Aliases:
AT3G60140, BETA GLUCOSIDASE 30, BGLU30, DARK INDUCIBLE 2, SENESCENCE-RELATED GENE 2, SRG2
Chromosome Location:
Chr.3: 22216540 - 22220824 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02197424_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340024.1 NP_001327679.1 5 - 6 626 147
NM_115877.4 NP_191573.1 5 - 6 626 147

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
aging
response to salt stress
glucosinolate catabolic process
hydrolase activity, hydrolyzing O-glycosyl compounds
beta-glucosidase activity
scopolin beta-glucosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0