This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02196912_g1
See other GCN2 GE Assays ›
Gene Symbol:
GCN2
Gene Name:
protein kinase family protein
Gene Aliases:
ARABIDOPSIS THALIANA GENERAL CONTROL NON-REPRESSIBLE 2, AT3G59410, ATGCN2, GENERAL CONTROL NON-REPRESSIBLE 2
Chromosome Location:
Chr.3: 21948851 - 21959410 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02196912_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203206.1 NP_001190135.1 17 - 18 2947 74
NM_001339969.1 NP_001326041.1 17 - 18 3814 74
NM_001339970.1 NP_001326042.1 16 - 17 2956 74
NM_001339971.1 NP_001326040.1 16 - 17 2727 74
NM_115803.3 NP_191500.2 16 - 17 2846 74
GenBank mRNA AJ459823.1 - 16 - 17 2494 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of cellular amino acid metabolic process
response to herbicide
peptidyl-serine phosphorylation
cellular response to amino acid starvation
cellular response to hydrogen peroxide
cellular response to benomyl
cellular stress response to acidic pH
tRNA binding
protein kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0