This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02194820_g1
See other HEMB2 GE Assays ›
Gene Symbol:
hemb2
Gene Name:
Aldolase superfamily protein
Gene Aliases:
AT1G44318, T18F15.10, T18F15_10
Chromosome Location:
Chr.1: 16831016 - 16834011 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02194820_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hemb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_103545.2 NP_175085.1 7 - 8 704 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
chlorophyll biosynthetic process
porphobilinogen synthase activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0