This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02192068_g1
See other LSP1 GE Assays ›
Gene Symbol:
LSP1
Gene Name:
Eukaryotic initiation factor 4E protein
Gene Aliases:
AT5G35620, EIF(ISO)4E, EIF4E2, EUKARYOTIC INITIATION FACTOR (ISO)4E, EUKARYOTIC INITIATION FACTOR (ISO)4E, EUKARYOTIC TRANSLATION INITATION FACTOR 4E2, LOSS OF SUSCEPTIBILITY TO POTYVIRUS 1, LSP, MJE4.8, MJE4_8, eIFiso4E, eukaryotic translation Initiation Factor isoform 4E
Chromosome Location:
Chr.5: 13824424 - 13826444 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02192068_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LSP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344112.1 NP_001332369.1 4 - 5 632 91
NM_122953.3 NP_198412.1 4 - 5 673 91
NM_203123.1 NP_974852.1 3 - 4 1039 91
GenBank mRNA AY054630.1 - 4 - 5 601 91
AY081514.1 - 4 - 5 560 91
AY086315.1 - 4 - 5 621 91
Y10547.1 - 587 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
response to virus
negative regulation of defense response to virus
RNA 7-methylguanosine cap binding
translation initiation factor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header