Assay Details
| Gene Symbol: AT3G57310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115591.2 | NP_191290.1 | 1 | 278 | 63 | |
More Information
Set Membership: |
|

AT3G57310| Gene Symbol: AT3G57310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115591.2 | NP_191290.1 | 1 | 278 | 63 | |
Set Membership: |
|