This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02189501_g1
See other TPI GE Assays ›
Gene Symbol:
TPI
Gene Name:
triosephosphate isomerase
Gene Aliases:
AT3G55440, ATCTIMC, CYTOSOLIC ISOFORM TRIOSE PHOSPHATE ISOMERASE, CYTOSOLIC TRIOSE PHOSPHATE ISOMERASE, CYTOTPI, TRIOSEPHOSPHATE ISOMERASE, triosephosphate isomerase
Chromosome Location:
Chr.3: 20553403 - 20556560 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02189501_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_115402.4 NP_191104.1 1 - 2 429 87
GenBank mRNA AF375426.1 - 1 - 2 132 87
AY074822.1 - 1 - 2 38 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
response to zinc ion
glycerol catabolic process
cellular carbohydrate metabolic process
glyceraldehyde-3-phosphate biosynthetic process
response to cadmium ion
triose-phosphate isomerase activity
copper ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0