This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02189167_g1
See other PDIL1-3 GE Assays ›
Gene Symbol:
PDIL1-3
Gene Name:
PDI-like 1-3
Gene Aliases:
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 1, AT3G54960, ATPDI1, ATPDIL1-3, PDI-like 1-3, PDI1, PROTEIN DISULFIDE ISOMERASE 1, T15C9.4
Chromosome Location:
Chr.3: 20363260 - 20366874 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02189167_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDIL1-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_115353.5 NP_191056.2 10 - 11 1573 67
GenBank mRNA AJ271376.1 - 10 - 11 1597 67
AY093115.1 - 10 - 11 1539 67
BT000172.1 - 10 - 11 1521 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to endoplasmic reticulum stress
cell redox homeostasis
protein disulfide isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0