This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02188790_m1
See other AT3G54360 GE Assays ›
Gene Symbol:
AT3G54360
Gene Name:
zinc ion binding protein
Gene Aliases:
AT3G54360, NCA1, NO CATALASE ACTIVITY 1
Chromosome Location:
Chr.3: 20128127 - 20131968 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02188790_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G54360
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001339663.1 NP_001327012.1 6 - 7 1177 69
NM_115296.3 NP_191004.1 6 - 7 1232 69
GenBank mRNA AF410273.1 - 6 - 7 1090 69
AY086065.1 - 6 - 7 1088 69
BT000620.1 - 5 - 6 1005 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
positive regulation of catalase activity
protein binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0