This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02187240_g1
See other SAC8 GE Assays ›
Gene Symbol:
SAC8
Gene Name:
SAC domain-containing protein 8
Gene Aliases:
AT3G51830, ATG5, SAC domain-containing protein 8, TRANSMEMBRANE PROTEIN G5P
Chromosome Location:
Chr.3: 19220116 - 19225280 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02187240_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SAC8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001339534.1 NP_001319727.1 5 - 6 629 68
NM_115042.3 NP_190751.2 5 - 6 624 68
GenBank mRNA AY080659.1 - 5 - 6 596 68
AY133741.1 - 5 - 6 508 68
AY227251.1 - 5 - 6 508 68
U72504.1 - 5 - 6 556 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol dephosphorylation
phosphoric ester hydrolase activity
phosphatidylinositol-4-phosphate phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0